Learn More
American Research Products Inc Mouse tumor necrosis factor li
Catalog No. 5000328430
Encompass
Mouse tumor necrosis factor-like ligand 1 (TL1) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28430 Supplier American Research Products Inc Supplier No. CSBE16311M
Mouse tumor necrosis factor-like ligand 1 (TL1) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich