Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU11538
Encompass_Preferred
Mouse Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) Wide-range ELISA Kit for detecting TRAIL in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50592. Gene ID: 22035. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8127