Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Receptor Superfamily, Member 17 (TNFRSF17) ELISA Kit
Supplier:  Biomatik Corporation EKU07972
Encompass_Preferred
Mouse Tumor Necrosis Factor Receptor Superfamily, Member 17 (TNFRSF17) ELISA Kit for detecting TNFRSF17 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.101ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88472. Gene ID: 21935. Target Synonyms: CD269; BCM; BCMA; B-cell maturation protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8710