Learn More
American Research Products Inc Mouse Tumor necrosis factor re
Catalog No. 5000326953
Encompass
Mouse Tumor necrosis factor receptor superfamily member 5 (CD40) ELISA kit from CUSABIO Detection Range 31 2 pg/ml-2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method
Catalog No. 50-003-26953 Supplier American Research Products Inc Supplier No. CSBEL004936MO
Mouse Tumor necrosis factor receptor superfamily member 5 (CD40) ELISA kit from CUSABIO Detection Range 31 2 pg/ml-2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method