Learn More
American Research Products Inc Mouse Tumor necrosis factor re
Catalog No. 5000328187
Encompass
Mouse Tumor necrosis factor receptor superfamily member 13B (TNFRSF13B) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28187 Supplier American Research Products Inc Supplier No. CSBEL023971MO
Mouse Tumor necrosis factor receptor superfamily member 13B (TNFRSF13B) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich