Learn More
American Research Products Inc Mouse Tumor necrosis factor re
Supplier:  American Research Products Inc CSBEL023972MO
Encompass
Mouse Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C) ELISA kit from CUSABIO Detection Range 39 pg/ml - 2500 pg/ml Sensitivity 9 75 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28251