Learn More
American Research Products Inc Mouse Tumor necrosis factor re
Supplier:  American Research Products Inc CSBEL023975MO
Encompass
Mouse Tumor necrosis factor receptor superfamily member 18 (TNFRSF18) ELISA kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27210