Learn More
American Research Products Inc Mouse Tumor necrosis factor re
Supplier:  American Research Products Inc CSBEL023984MO
Encompass
Mouse Tumor necrosis factor receptor superfamily member 9 (TNFRSF9) ELISA kit from CUSABIO Detection Range 15 6 pg/ml - 1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27799