Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Receptor Superfamily, Member 5 (CD40) ELISA Kit
Supplier:  Biomatik Corporation EKU07978
Encompass_Preferred
Mouse Tumor Necrosis Factor Receptor Superfamily, Member 5 (CD40) ELISA Kit for detecting CD40 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27512. Gene ID: 21939. Target Synonyms: TNFRSF5; CDW40; Bp50; P50; B-cell surface antigen CD40; CD40L receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7213