Learn More
American Research Products Inc Mouse Tumor necrosis factor so
Catalog No. 5000326519
Encompass
Mouse Tumor necrosis factor soluble receptor II TNFsR-II ELISA KIT from CUSABIO Detection Range 7 8125 pg/mL-500 pg/mL Sensitivity 1 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26519 Supplier American Research Products Inc Supplier No. CSBE04739M
Mouse Tumor necrosis factor soluble receptor II TNFsR-II ELISA KIT from CUSABIO Detection Range 7 8125 pg/mL-500 pg/mL Sensitivity 1 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich