Learn More
Biomatik Corporation Mouse Twist Transcription Factor (TWIST) ELISA Kit
Supplier:  Biomatik Corporation EKU09396
Encompass_Preferred
Mouse Twist Transcription Factor (TWIST) ELISA Kit for detecting TWIST in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26687. Gene ID: 22160. Target Synonyms: ACS3; BPES2; BPES3; SCS; Class A basic helix-loop-helix protein 38; Acrocephalosyndactyly 3; Saethre-Chotzen Syndrome; Blepharophimosis,Epicanthus Inversus And Ptosis 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7140