Learn More
Sino Biological Anti-Thioredoxin/TRX Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50822RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Thioredoxin / TXN / SASP (rM Thioredoxin / TXN / SASP; Catalog#50822-MNAE; P10639; Met1-Val105). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3067