Learn More
Sino Biological Anti-Thioredoxin/TRX Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50822T50100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Thioredoxin/TXN/SASP (rM Thioredoxin/TXN/SASP; Catalog#50822-MNAE; P10639; Met1-Val105). Thioredoxin/TXN/SASP specific IgG was purified by Mouse Thioredoxin/TXN/SASP affinity chromatography.
Catalog No. 50-255-3066