Learn More
Biomatik Corporation Mouse Ubiquinol Cytochrome C Reductase Core Protein II (UQCRC2) ELISA Kit
Supplier:  Biomatik Corporation EKU08011
Encompass_Preferred
Mouse Ubiquinol Cytochrome C Reductase Core Protein II (UQCRC2) ELISA Kit for detecting UQCRC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9DB77. Gene ID: 67003. Target Synonyms: QCR2; UQCR2; Cytochrome b-c1 complex subunit 2, mitochondrial; Complex III subunit 2; Core protein II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5535