Learn More
Biomatik Corporation Mouse Ubiquitin Carboxyl Terminal Hydrolase L4 (UCHL4) ELISA Kit
Supplier:  Biomatik Corporation EKU11506
Encompass_Preferred
Mouse Ubiquitin Carboxyl Terminal Hydrolase L4 (UCHL4) ELISA Kit for detecting UCHL4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58321. Gene ID: 93841. Target Synonyms: Ubiquitin thioesterase L4; Ubiquitin carboxyl-terminal hydrolase isozyme L4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6576