Learn More
American Research Products Inc Mouse Ubiquitin like protein I
Catalog No. 5000327065
Encompass
Mouse Ubiquitin-like protein ISG15 (ISG15) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27065 Supplier American Research Products Inc Supplier No. CSBEL011843MO
Mouse Ubiquitin-like protein ISG15 (ISG15) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich