Learn More
Biomatik Corporation Mouse Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit
Supplier:  Biomatik Corporation EKU12362
Encompass_Preferred
Mouse Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit for detecting UBE3A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.277ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08759. Gene ID: 22215. Target Synonyms: ANCR; AS; E6-AP; EPVE6AP; HPVE6A; Papilloma Virus E6-Associated,Angelman Syndrome; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7977