Learn More
Sino Biological Anti-UCHL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50690RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse UCHL1 (rM UCHL1; Catalog#50690-M07E; Q9R0P9; Gln 2-Ala 223). UCHL1 specific IgG was purified by Mouse UCHL1 affinity chromatography.
Catalog No. 50-241-2595