Learn More
World Precision Instrument Mouse Ultra Precise Dual Stereotaxic Instrument
SDP
Catalog No. 502539123
Encompass
Our Smallest and Lowest Priced Stereotaxic Instrument!
The WPI Mouse Stereotaxic Instruments was specifically designed for use with knock-out and transgenic mice. It is our smallest and most economic stereotaxic instrument. The WPI Mouse Stereotaxic Instruments includes a 100 micron, 3-axes left-hand manipulator arm, mouse snout clamp and tooth bar, dual sided ear bars, and a corner clamp probe holder. Available as Dual, Ultra-Precise, Digital or Motorized.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
Catalog No. 50-253-9123 Supplier World Precision Instrument Supplier No. 505321
Add to Cart
Add to Cart
Request a Quote
Our Smallest and Lowest Priced Stereotaxic Instrument!
The WPI Mouse Stereotaxic Instruments was specifically designed for use with knock-out and transgenic mice. It is our smallest and most economic stereotaxic instrument. The WPI Mouse Stereotaxic Instruments includes a 100 micron, 3-axes left-hand manipulator arm, mouse snout clamp and tooth bar, dual sided ear bars, and a corner clamp probe holder. Available as Dual, Ultra-Precise, Digital or Motorized.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.