Learn More
American Research Products Inc Mouse UndercarboXylated Osteoc
Catalog No. 5000328365
Encompass
Mouse UndercarboXylated Osteocalcin (ucOC) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28365 Supplier American Research Products Inc Supplier No. CSBE17681M
Mouse UndercarboXylated Osteocalcin (ucOC) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich