Learn More
Biomatik Corporation Mouse Upregulator Of Cell Proliferation (URGCP) ELISA Kit
Supplier:  Biomatik Corporation EKU08041
Encompass_Preferred
Mouse Upregulator Of Cell Proliferation (URGCP) ELISA Kit for detecting URGCP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5NCI0. Gene ID: 72046. Target Synonyms: URG4; Up Regulated Gene 4; HBV X protein up-regulated gene 4 protein; HBxAg up-regulated gene 4 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5530