Learn More
Biomatik Corporation Mouse Uridine Phosphorylase 1 (UPP1) ELISA Kit
Supplier:  Biomatik Corporation EKU09087
Encompass_Preferred
Mouse Uridine Phosphorylase 1 (UPP1) ELISA Kit for detecting UPP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52624. Gene ID: 22271. Target Synonyms: UDRPASE; UP; UPASE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7465