Learn More
Crystal Chem Inc Mouse Urinary NAG Assay Kit, 96-tests
SDP
Catalog No. 501947703
Encompass_Preferred
The Mouse Urinary of NAG) Assay is used to quantify levels of NAG in mouse urine using only a 10 µL sample volume.Highlights: Kit uses sample size of only 10 µL, ideal for mouse applications. Excellent correlation to ELISA method. Less than 15 minute procedure. Can be run manually or fully automated on a wide range of instruments. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7703 Supplier Crystal Chem Inc Supplier No. 80400
The Mouse Urinary of NAG) Assay is used to quantify levels of NAG in mouse urine using only a 10 µL sample volume.Highlights: Kit uses sample size of only 10 µL, ideal for mouse applications. Excellent correlation to ELISA method. Less than 15 minute procedure. Can be run manually or fully automated on a wide range of instruments. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.