Learn More
Biomatik Corporation Mouse Urocortin (UCN) ELISA Kit
Supplier:  Biomatik Corporation EKU10253
Encompass_Preferred
Mouse Urocortin (UCN) ELISA Kit for detecting UCN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81615. Gene ID: 22226. Target Synonyms: UI; UROC; Urocortin 1; UCN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8358