Learn More
American Research Products Inc Mouse urokinase plasminogen ac
Catalog No. 5000326473
Encompass
Mouse urokinase plasminogen actiVator uPA ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26473 Supplier American Research Products Inc Supplier No. CSBE07369M
Mouse urokinase plasminogen actiVator uPA ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich