Learn More
Biomatik Corporation Mouse Urotensin 2 (UST2) ELISA Kit
Supplier:  Biomatik Corporation EKU08063
Encompass_Preferred
Mouse Urotensin 2 (UST2) ELISA Kit for detecting UST2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.43pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9QZQ3. Gene ID: 24111. Target Synonyms: UII; U-II; UCN2; PRO1068; Urotensin-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6922