Learn More
XpressBio Mouse Anti-HBsAg ELISA Kit
SDP
Supplier:  XpressBio IM325
Encompass_Preferred
Quantitative ELISA assay developed to specifically measure the degree of immunity against a specific range of infectious targets. The microtiter wells coated with recombinant HBsAg (adw and adr), areexposed to test specimens, which may contain mouse IgG reactive antibodies.
Catalog No. 50-171-7662