Learn More
American Research Products Inc Mouse Vascular cell adhesion m
Catalog No. 5000326524
Encompass
Mouse Vascular cell adhesion molecule 1 VCAM-1 ELISA kit from CUSABIO Detection Range 39 062 pg/mL-2500 pg/mL Sensitivity 29 649 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26524 Supplier American Research Products Inc Supplier No. CSBE04754M
Mouse Vascular cell adhesion molecule 1 VCAM-1 ELISA kit from CUSABIO Detection Range 39 062 pg/mL-2500 pg/mL Sensitivity 29 649 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich