Learn More
Biomatik Corporation Mouse Vascular Cell Adhesion Molecule 1, Vcam-1 Elisa Kit
Supplier:  Biomatik Corporation EKC31435
Encompass_Preferred
Mouse Vascular Cell Adhesion Molecule 1, Vcam-1 Elisa Kit for detecting Vascular Cell Adhesion Molecule 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 39.062-2500pg/ml. Sensitivity: 29.649pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29533; Vcam1. Target Synonyms: Vcam1; Vcam-1; Vascular cell adhesion protein 1; V-CAM 1; VCAM-1; CD antigen CD106. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6647