Learn More
American Research Products Inc Mouse Vascular endothelial cel
Supplier:  American Research Products Inc CSBE04762M
Encompass
Mouse Vascular endothelial cell growth factor receptor 1 VEGFR-1/Flt1 ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29071