Learn More
American Research Products Inc Mouse Vascular Endothelial cel
Catalog No. 5000329922
Encompass
Mouse Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 3 906 pg/mL-250 pg/mL Sensitivity 0 857 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29922 Supplier American Research Products Inc Supplier No. CSBE04756M
Mouse Vascular Endothelial cell Growth Factor VEGF ELISA KIT from CUSABIO Detection Range 3 906 pg/mL-250 pg/mL Sensitivity 0 857 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich