Learn More
American Research Products Inc Mouse Vascular endothelial cel
Supplier:  American Research Products Inc CSBE04766M
Encompass
Mouse Vascular endothelial cell growth factor receptor 3 (VEGFR-3/Flt-4) ELISA kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26312