Learn More
American Research Products Inc Mouse Vascular endothelial cel
Catalog No. 5000326521
Encompass
Mouse Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26521 Supplier American Research Products Inc Supplier No. CSBE04764M
Mouse Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich