Learn More
American Research Products Inc Mouse Vascular Endothelial cel
Supplier:  American Research Products Inc CSBE07361M
Encompass
Mouse Vascular Endothelial cell Growth Factor C VEGF-C ELISA kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26469