Learn More
American Research Products Inc Mouse Vascular Endothelial cel
Supplier:  American Research Products Inc CSBE07357M
Encompass
Mouse Vascular Endothelial cell Growth Factor D VEGF-D ELISA kit from CUSABIO Detection Range 0 312 pg/mL-20 pg/mL Sensitivity 0 078 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26665