Learn More
American Research Products Inc Mouse Vascular Endothelial cel
Supplier:  American Research Products Inc CSBE07365M
Encompass
Mouse Vascular Endothelial cell Growth Factor B VEGF-B ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26471