Learn More
Biomatik Corporation Mouse Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit
Supplier:  Biomatik Corporation EKC31436
Encompass_Preferred
Mouse Vascular Endothelial Cell Growth Factor, Vegf Elisa Kit for detecting Vascular Endothelial Growth Factor A in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.906-250pg/ml. Sensitivity: 0.857pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q00731; Vegfa. Target Synonyms: Vegfa; Vegf; Vascular endothelial growth factor A; VEGF-A; Vascular permeability factor; VPF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6645