Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor 121 (VEGF121) ELISA Kit
Supplier:  Biomatik Corporation EKU11513
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor 121 (VEGF121) ELISA Kit for detecting VEGF121 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00731. Gene ID: 22339. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5444