Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) CLIA Kit
Supplier:  Biomatik Corporation EKU08946
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) CLIA Kit for detecting VEGFR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P35969. Gene ID: 14254. Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8587