Learn More
Proteintech Group Inc Mouse VCAM-1/CD106 ELISA
SDP
Supplier:  Proteintech Group Inc KE1003896T
Encompass_Preferred
Vascular cell adhesion molecule 1 (VCAM-1), also known as CD106, is a transmembrane glycoprotein belonging to the immunoglobulin gene superfamily. VCAM-1 is expressed by cytokine-activated endothelium, interacts with integrin VLA4 (α4β1) present on the surface of leukocytes, and mediates both adhesion and signal transduction. The VCAM-1/integrin VLA4 (α4β1) interaction may play a pathophysiologic role both in immune responses and in leukocyte emigration to sites of inflammation. VCAM-1 is also expressed either constitutively or inducibly in a variety of other cell types, including vascular smooth muscle cells, differentiating skeletal muscle cells, renal and neural epithelial cells, macrophages (Kupffer cells), dendritic cells, and bone marrow stromal cells. Soluble VCAM-1 (sVCAM-1) has been found in addition to membrane-bound VCAM-1.
Catalog No. 50-197-4162