Learn More
Sino Biological Mouse VCAM1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 202919T44100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse VCAM-1 / CD106 (rM VCAM-1 / CD106; Catalog#50163-M08H; NP_035823.3; Met 1-Glu 698). VCAM-1 / CD106 specific IgG was purified by Mouse VCAM-1 / CD106 affinity chromatography.
Catalog No. 50-237-4297