Learn More
Sino Biological Mouse VCAM1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50163M08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 698) of mouse VCAM1 (NP_035823.3) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3161