Learn More
Sino Biological Anti-Vitamin D Receptor/VDR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51106T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Vitamin D Receptor / VDR (rM Vitamin D Receptor / VDR; Catalog#51106-M08B; P48281; Met1-Ser422). Vitamin D Receptor / VDR specific IgG was purified by Mouse Vitamin D Receptor / VDR affinity chromatography.
Catalog No. 50-241-2336