Learn More
FUJIFILM BIOSCIENCES INC Mouse VEGF-165, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 200341MG
Encompass_Preferred
Vascular endothelial growth factor A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells.  VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways.  VEGF-A has several alternatively spliced isoforms, with VEGF-165 being the most abundant.  The VEGF-165 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell proliferation and angiogenesis.
Catalog No. 50-990-864