Learn More
Kingfisher Biotech Inc Mouse VEGF-A Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc KP0742M100
Encompass_Preferred
The Mouse VEGF-A polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Mouse VEGF-A ELISA. The Mouse VEGF-A antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine VEGF-A - Strong) (Canine VEGF-A - Strong) (Caprine VEGF-A - Strong) (Chicken/Turkey VEGF-A - Weak) (Equine VEGF-A - Strong) (Feline VEGF-A - Strong) (Human/Monkey VEGF-A - Strong) (Mouse VEGF-A - Strong) (Ovine VEGF-A - Strong) (Rabbit VEGF-A - Strong) (Rat VEGF-A - Strong) (Swine VEGF-A - Strong) (Zebrafish VEGF-A - None). For research use only. Made in the USA
Catalog No. 50-233-2332