Learn More
Raybiotech Inc Mouse VEGF-B ELISA Kit
Supplier:  Raybiotech Inc ELMVEGFB1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse 6Ckine, Activin A, ADAMTS1, Adiponectin, ANG-3, ANGPTL3, Artemin, CCL28, CD36, Chordin, CRP, E-Cadherin, Epigen, Epiregulin, Fas, Galectin-7, gp130, Granzyme B, Gremlin, IFN-gamma R1, IL-17B, IL-17B R, IL-22, MIP-1 beta, MMP-2, MMP-3, MMP-10, PDGF-AA, Persephin, sFRP-3, Shh-N, SLAM, TCK-1, TECK, TGF beta-1, TRANCE, TremL1, TWEAK, VEGF-R2.
Catalog No. 50-195-0160