Learn More
Sino Biological Anti-VEGF164 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50159RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse VVEGF164/VEGFA (Catalog#50159-MNAB; Q00731-2; Met1-Arg190). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2577