Learn More
Raybiotech Inc Mouse VEGFR3 ELISA Kit
Supplier:  Raybiotech Inc ELMVEGFR31
Encompass_Preferred
For Cell Culture Supernatants, EDTA and/or Citrate treated plasma, and Serum samples. Heparin is not recommended. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Mouse Vascular endothelial growth factor receptor 3.
Catalog No. 50-195-0164