Learn More
BIOHIPPO Mouse very Microsample
SDP
Catalog No. 5000315290
Encompass
Mouse very Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of very in serum plasma cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 g/mL with a sensitivity of 0 98 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VLDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VLDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VLDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15290 Supplier BIOHIPPO Supplier No. BHE15212694
Mouse very Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of very in serum plasma cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 g/mL with a sensitivity of 0 98 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse VLDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse VLDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse VLDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color