Learn More
Biomatik Corporation Mouse V-Ets Erythroblastosis Virus E26 Oncogene Homolog 1 (ETS1) ELISA Kit
Supplier:  Biomatik Corporation EKU08135
Encompass_Preferred
Mouse V-Ets Erythroblastosis Virus E26 Oncogene Homolog 1 (ETS1) ELISA Kit for detecting ETS1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27577. Gene ID: 23871. Target Synonyms: EWSR2; p54; Protein C-ets-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6989